Learn More
R&D Systems™ Z-Leu-Leu-Leu-B(OH)2 (MG-262)
Highly potent and selective cell-permeable inhibitor of the proteasome
Supplier:  R&D Systems™ I120200
 View more versions of this product
Catalog No. I120200
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
  • This peptide boronic acid is structurally similar to the peptide aldehyde MG-132 and also inhibits the chymotrypsin-like peptidase activity of the proteasome, but with much higher potency
  • These inhibitors are reversible and also inhibit the activity of calpains and cathepsins
  • Purity >95% by HPLC
  • Structure confirmed by 1H-NMR
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Ubiquitinase Inhibitor
Proteasome
200 μg
491.44
Product Suggestions
Product Suggestions