Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PS 1145 dihydrochloride
Selective IKK inhibitor; orally active
$286.52 - $1177.40
Specifications
Purity | >98% |
---|---|
Product Type | PS 1145 dihydrochloride |
Molecular Weight (g/mol) | 395.67 |
Inhibitors | PS 1145 dihydrochloride |
Description
Selective IκB kinase (IKK) inhibitor (IC50 = 100 nM). Has no effect on PKA, PKC and CKII activity. Inhibits IκB-α phosphorylation and NFκB activation in HeLa cells. Inhibits NFκB-mediated proinflammatory gene expression in human airway epithelial cells. Enhances TNF-α-induced apoptosis in a multiple myeloma cell line. Attenuates LPS-induced TNF-α production in vivo. Orally active.Specifications
>98% | |
PS 1145 dihydrochloride | |
395.67 | |
PS 1145 dihydrochloride |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title