Learn More
Cole-Parmer™ Oakton™ Portable Turbidity Meter Kit
Provides accurate results with simple calibration and testing
Supplier:  Cole-Parmer™ 3563510
 View more versions of this product
Catalog No. 13-000-101
Explore available promotions
Add to Cart
Description
Description
  • Auto-ranging from 0 to 1000 NTU (FNU)
  • Stores up to 200 sets of data
  • Suitable for field use
  • Meets IP67 water-resistant requirements
Specifications
Specifications
Turbidity Meter
Color TFT
Measuring, Calibration, Parameter Setup, Data Recall
≤ ±2% of reading stray light
0 to 1000 NTU
Rechargeable Lithium-ion Battery
2 Years
White Light
Variable
IP67
0.01/0.1/1 NTU
20 Hours
EPA 180.1 method
Product Suggestions
Product Suggestions