Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Polypropylene Cryo Cube Box
SDPSCI_ED

Storage boxes that can be used with multiple tube sizes.

$31.50 - $39.30

Specifications

Closure Material Polypropylene
Color Blue
Material Polypropylene
Shape Square
Type Cryo Box
View More Specs

Products 2
Catalog Number Mfr. No. Holds Price Quantity & Availability  
Catalog Number Mfr. No. Holds Price Quantity & Availability  
S01151
SDPSCI_ED
View Documents
United Scientific™
66501
81 Tubes
Pack of 4 for $31.50
Add to Cart
 
S01152
SDPSCI_ED
View Documents
United Scientific™
66502
100 Tubes
Pack of 4 for $39.30
Add to Cart
 
Description

Description

  • Manufactured of polypropylene, these autoclavable boxes are stackable for sample storage in 1.5 to 2.0 mL microtubes and 1.0 to 2.0 mL cryogenic vials
  • Boxes can be used at temperatures from -90°C to +121°C and are autoclavable
  • Boxes feature a hinged lid connected to the base and a locking system to ensure secure closure
  • A molded alphanumeric grid allows for easy identification of samples
  • Dimensions of cryo boxes are designed to be compatible with vertical, horizontal, and drawers style freezer racks
  • Each pack includes two yellow boxes and two blue boxes
Specifications

Specifications

Polypropylene
Polypropylene
Cryo Box
Blue
Square
SDS
Documents

Documents

Product Certifications

Warning: Cancer and reproductive harm

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit