Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Glycohemoglobin Reagents
Specifications
Certifications/Compliance | CE Marked |
---|---|
Type | Glycohemoglobin Reagent |
For Use With (Application) | Quantitative determination of Glycohemoglobin (HbA1) in Blood by cation exchange resin, test is to be used to monitor long-term Glucose control in diabetes mellitus |
Includes | 1 x 120mL Bottle 8mg/mL cation-exchange resin in a Borate Buffer, pH 6.9, 1 x 30mL bottle Glycohemoglobin Lysing resin, 10mM Potassium Cyanide, Surfactant added, 40 Serum separators |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-666-293
|
Pointe Scientific
G754040 |
1 x 120mL Resin, 1 x 30mL Lysing, 40 Serum separators | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Specifications
CE Marked | |
Quantitative determination of Glycohemoglobin (HbA1) in Blood by cation exchange resin, test is to be used to monitor long-term Glucose control in diabetes mellitus | |
Liquid |
Glycohemoglobin Reagent | |
1 x 120mL Bottle 8mg/mL cation-exchange resin in a Borate Buffer, pH 6.9, 1 x 30mL bottle Glycohemoglobin Lysing resin, 10mM Potassium Cyanide, Surfactant added, 40 Serum separators |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title