Learn More
Pointe Scientific Glycohemoglobin Reagents
Supplier: Pointe Scientific G754040
This item is not returnable.
View return policy
Specifications
CE Marked | |
Quantitative determination of Glycohemoglobin (HbA1) in Blood by cation exchange resin, test is to be used to monitor long-term Glucose control in diabetes mellitus | |
1 x 120mL Resin, 1 x 30mL Lysing, 40 Serum separators |
Glycohemoglobin Reagent | |
1 x 120mL Bottle 8mg/mL cation-exchange resin in a Borate Buffer, pH 6.9, 1 x 30mL bottle Glycohemoglobin Lysing resin, 10mM Potassium Cyanide, Surfactant added, 40 Serum separators | |
Liquid |