Learn More
Pointe Scientific Detergent Reagents for Hitachi™ Analyzers
Detergent solutions designed specifically for Hitachi Analyzers
Supplier: Pointe Scientific HI790DET
Includes: Sodium hydroxide (1 to 5%); Ethanolamine (10 to 20%); EDTA (5 to 10%); Water (50 to 75%); Polyoxyethylene lauryl ether (1 to 5%); Emulgator K-30 (Paraffin oils and Kerosene) (1 to 5%)
Description
- Liquid format
For use with Hitachi analyzers
Specifications
CE Marked | |
Hitachi 704, 717, 747, 902 analyzer | |
Liquid |
Detergent | |
1 x 1 L |