Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Detergent Reagents for Hitachi™ Analyzers
Detergent solutions designed specifically for Hitachi Analyzers
$260.00
Specifications
Certifications/Compliance | CE Marked |
---|---|
Type | Detergent |
Form | Liquid |
Includes: Sodium hydroxide (1 to 5%); Ethanolamine (10 to 20%); EDTA (5 to 10%); Water (50 to 75%); Polyoxyethylene lauryl ether (1 to 5%); Emulgator K-30 (Paraffin oils and Kerosene) (1 to 5%)
Catalog Number | Mfr. No. | Quantity | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | For Use With (Equipment) | Price | Quantity & Availability | ||||
23-666-251
|
Pointe Scientific
HI790DET |
1 x 1 L | Hitachi 704, 717, 747, 902 analyzer |
Each for $260.00
|
|
||||
23-666-252
|
Pointe Scientific
HI990DET |
14 x 100 mL | Hitachi 911, 912, 914 analyzer | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Liquid format
For use with Hitachi analyzers
Specifications
CE Marked | |
Detergent | |
Liquid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title