Learn More
Pointe Scientific BUN Reagents
Supplier: Pointe Scientific HB9521020
This item is not returnable.
View return policy
Specifications
CE Marked | |
Urease (Jack Bean) >15000U/L, GLDH (Bovine) >200U/L, ADP >0.6mM, alpha-Ketoglutarate 3.6mM, NADH >0.28mM, Buffer pH 7.8 ± 0.1, Stabilizers, Sodium Azide 0.25% as preservative | |
Hitachi 717 analyzers | |
Liquid |
BUN Reagent Kit | |
Quantitative determination of Urea Nitrogen in Serum | |
8 x 100mL R1, 4 x 50mL R2 |