Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific BUN Reagents
$588.00
Specifications
Certifications/Compliance | CE Marked |
---|---|
For Use With (Application) | Quantitative determination of Urea Nitrogen in Serum |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Formulation | For Use With (Equipment) | Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Formulation | For Use With (Equipment) | Type | Price | Quantity & Availability | ||
23-666-156
|
Pointe Scientific
HB9521020 |
8 x 100mL R1, 4 x 50mL R2 | Urease (Jack Bean) >15000U/L, GLDH (Bovine) >200U/L, ADP >0.6mM, alpha-Ketoglutarate 3.6mM, NADH >0.28mM, Buffer pH 7.8 ± 0.1, Stabilizers, Sodium Azide 0.25% as preservative | Hitachi 717 analyzers | BUN Reagent Kit |
Pack of 12 for $588.00
|
|
||
23-666-160
|
Pointe Scientific
HB452R1 |
5 x 600 R1mL | Ketoglutaric Acid 0.2%, Sodium Azide 0.095% | Hitachi 747 analyzers | BUN R1 | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
23-666-161
|
Pointe Scientific
HB452R2 |
5 x 300 R2mL | Ketoglutaric Acid 0.2%, Sodium Azide 1% | Hitachi 747 analyzers | BUN R2 | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
23-666-154
|
Pointe Scientific
HB752510 |
8 x 50mL R1, 2 x 50mL R2 | Urease (Jack Bean) >15000U/L, GLDH (Bovine) >200U/L, ADP >0.6mM, alpha-Ketoglutarate 3.6mM, NADH >0.28mM, Buffer pH 7.8 ± 0.1, Stabilizers, Sodium Azide 0.25% as preservative | Hitachi 704 analyzers | BUN Reagent Kit | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
Specifications
CE Marked | |
Quantitative determination of Urea Nitrogen in Serum | |
Liquid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title