Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Acid Phosphatase Reagents
$231.00 - $274.00
Specifications
Certifications/Compliance | CE Marked |
---|---|
Type | Acid Phosphatase Reagent Kit |
Formulation | Acid Phosphatase reagent: alpha-naphthylphosphate 3mM, Fast Red TR 1mM, Citric Acid 20mM, Sodium Citrate 60mM pH 5.3 ± 0.1, L-Tartrate reagent: Sodium L Tartrate 2M, Citric Acid 70mM, Sodium Citrate 10mM pH 5.3 ± 0.1, A |
For Use With (Application) | Quantitative determination of Acid Phosphatase in Serum |
Form | Dry Powder |
Catalog Number | Mfr. No. | Quantity | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | For Use With (Equipment) | Price | Quantity & Availability | ||||
23-666-059
|
Pointe Scientific
A7503100 |
10 x 10mL Reagent, 1 x 5mL L-Tartrate, 1 x 15mL Acetate Buffer | Hitachi analyzer |
Pack of 10 for $274.00
|
|
||||
23-666-061
|
Pointe Scientific
HA70360 |
6 x 10 mL | Hitachi 704 automatic analyzer |
Pack of 6 for $231.00
|
|
||||
Description
Reagent Set 1 includes Acid Phosphatase Reagent (10 x 10mL), L-Tartrate Reagent (5mL), and Acetate Buffer (15mL). Reagent Kit HA703-60 is used with Hitachi 704, 717.
Specifications
CE Marked | |
Acid Phosphatase reagent: alpha-naphthylphosphate 3mM, Fast Red TR 1mM, Citric Acid 20mM, Sodium Citrate 60mM pH 5.3 ± 0.1, L-Tartrate reagent: Sodium L Tartrate 2M, Citric Acid 70mM, Sodium Citrate 10mM pH 5.3 ± 0.1, A | |
Dry Powder |
Acid Phosphatase Reagent Kit | |
Quantitative determination of Acid Phosphatase in Serum |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title