Learn More
Pointe Scientific Acid Phosphatase Reagents
SDP
Supplier:  Pointe Scientific HA70360
 View more versions of this product
Catalog No. 23-666-061
Add to Cart
This item is not returnable. View return policy
Description
Description
Reagent Set 1 includes Acid Phosphatase Reagent (10 x 10mL), L-Tartrate Reagent (5mL), and Acetate Buffer (15mL). Reagent Kit HA703-60 is used with Hitachi 704, 717.
Specifications
Specifications
CE Marked
Acid Phosphatase reagent: alpha-naphthylphosphate 3mM, Fast Red TR 1mM, Citric Acid 20mM, Sodium Citrate 60mM pH 5.3 ± 0.1, L-Tartrate reagent: Sodium L Tartrate 2M, Citric Acid 70mM, Sodium Citrate 10mM pH 5.3 ± 0.1, A
Hitachi 704 automatic analyzer
Dry Powder
Acid Phosphatase Reagent Kit
Quantitative determination of Acid Phosphatase in Serum
6 x 10 mL