Learn More
Pig Kidney
Ideal for showing organ structure.
Catalog Number | Mfr. No. | Injection Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Injection Type | Price | Quantity & Availability | |||||
S1452S
|
Ranaco Corporation
140.1453.1 |
Plain |
Pack for $7.20
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S1456S
|
N/A | Single |
Pack
for $4.53
Save 48% Reg : |
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S9208
|
Ranaco Corporation
140.1461.1 |
Double |
Pack for $16.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S9210
|
Ranaco Corporation
140.1465.1 |
Triple |
Pack for $16.75
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Show kidney structure by dissecting these superior specimens.
INJECTION KEY:
- Plain: No color injections
- Single: Renal artery colored with red latex
- Double: Renal artery colored red, renal vein colored blue
- Triple: Renal artery colored red, renal vein colored blue, ureter and renal pelvis colored yellow
Specifications
Pig Kidney | |
Formaldehyde |