Learn More
Pig Kidney
Ideal for showing organ structure.
Supplier: Ranaco Corporation 140.1461.1
Description
Show kidney structure by dissecting these superior specimens.
INJECTION KEY:
- Plain: No color injections
- Single: Renal artery colored with red latex
- Double: Renal artery colored red, renal vein colored blue
- Triple: Renal artery colored red, renal vein colored blue, ureter and renal pelvis colored yellow
Specifications
Pig Kidney | |
Formaldehyde | |
Double | |
50/Cs. |