Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SKY-Z Series Horizontal Wind Turbines
Explore wind energy in this compact, innovative turbine design.
Specifications
Grade | For ages 12+ when using the recitifcation system but 8+ when using just the basic foundation model |
---|---|
For Use With (Application) | Turbine creates alternating current power with the ability to rectify power to direct current |
Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
S04700
|
Picoturbine International
PWH1 |
Turbine creates alternating current power with the ability to rectify power to direct current | SKY-Z Horizontal Wind Turbine | 34 oz. | 0.9 kg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
Description
Explore wind energy in this compact, innovative turbine design.
- Creates alternating current power
- Built-in LED light indicates generated power
- Can also accommodate different blade sizes and six-blade hub
- Dimensions (blade to floor × nose to floor × base): 26 × 18 × 6.2 in. (660 × 457 × 157.5mm)
- Blades and hubs also available separately
SKY-Z Horizontal Wind Turbine
- With three 8 in. (203mm) blades within three-blade hub with pitch angles, four coils and four magnets to build and customize generator
- With six 8 in. (203mm) blades within six-blade hub with pitch angles, four coils and four magnets to build and customize generator
- With rectifier to convert AC to DC power, with four types of rectification experiments
Specifications
For ages 12+ when using the recitifcation system but 8+ when using just the basic foundation model | |
Turbine creates alternating current power with the ability to rectify power to direct current |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title