Learn More
SKY-Z Series Horizontal Wind Turbines
Explore wind energy in this compact, innovative turbine design.
Supplier: Picoturbine International PWH1
This item is not returnable.
View return policy
Description
Explore wind energy in this compact, innovative turbine design.
- Creates alternating current power
- Built-in LED light indicates generated power
- Can also accommodate different blade sizes and six-blade hub
- Dimensions (blade to floor × nose to floor × base): 26 × 18 × 6.2 in. (660 × 457 × 157.5mm)
- Blades and hubs also available separately
SKY-Z Horizontal Wind Turbine
- With three 8 in. (203mm) blades within three-blade hub with pitch angles, four coils and four magnets to build and customize generator
- With six 8 in. (203mm) blades within six-blade hub with pitch angles, four coils and four magnets to build and customize generator
- With rectifier to convert AC to DC power, with four types of rectification experiments
Specifications
0.9 kg | |
SKY-Z Horizontal Wind Turbine | |
4 Coils, 4 magnets for generator, 3 blades and 3 blade hub |
For ages 12+ when using the recitifcation system but 8+ when using just the basic foundation model | |
34 oz. | |
Turbine creates alternating current power with the ability to rectify power to direct current |