Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture, pH 5.2±0.3, Liq.), Fisher BioReagents
Specifications
Chemical Name or Material | Phenol/Chloroform/Isoamyl Alcohol |
---|---|
Boiling Point | 96°C |
Molecular Formula | CHCl3 |
MDL Number | MFCD00000826 |
Merck Index | 15, 7354 |
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
BP1753I-400
|
400 mL | Coated Amber Glass/Poison Pack | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | |||||
Description
This mixture is used for extracting protein from nucleic acid preparations.Specifications
Phenol/Chloroform/Isoamyl Alcohol | |
CHCl3 | |
15, 7354 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
trichloromethane | |
≥99.0% | |
Coated Amber Glass/Poison Pack | |
Liquid |
96°C | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
ClC(Cl)Cl | |
119.37 | |
Pass Test | |
5.2 |
Safety and Handling
DOTInformation : DOT Class 6.1, : Poison
Recommended Storage : 4°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title