Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture, pH 5.2±0.3, Liq.), Fisher BioReagents
Description
This mixture is used for extracting protein from nucleic acid preparations.Specifications
Phenol/Chloroform/Isoamyl Alcohol | |
CHCl3 | |
15, 7354 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
trichloromethane | |
≥99.0% | |
Coated Amber Glass/Poison Pack | |
400 mL |
96°C | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
ClC(Cl)Cl | |
119.37 | |
Pass Test | |
5.2 | |
Liquid |
Safety and Handling
DOTInformation : DOT Class 6.1, : Poison
Recommended Storage : 4°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction