Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BrandTech™ BRAND™ PCR 12-Strip Caps
Manufactured from polypropyle without lubricants or chemical additives that can contaminate reagents and samples.PCR 12-Strip Caps are autoclavable and RNase-, DNase-, and DNA-free. Caps are domed and available in a variety of colors for sample identification.
$133.00
Specifications
For Use With (Equipment) | PCR tubes |
---|---|
Shape | Domed |
Material | Polypropylene |
Dnase Free | Yes |
RNase Free | Yes |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
13-882-312
|
BrandTech™ BRAND™
781290 |
Clear |
Pack of 125 for $133.00
|
|
|||||
13-882-313
|
BrandTech™ BRAND™
781291 |
Rose | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
13-882-314
|
BrandTech™ BRAND™
781292 |
Yellow |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||||
13-882-316
|
BrandTech™ BRAND™
781294 |
Blue | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples.- Autoclavable
- RNase-, DNase-, and DNA-free
- All 12-Strip caps are domed and available in a variety of colors for sample identification
Specifications
PCR tubes | |
Polypropylene | |
Yes | |
Tube Strip Cap |
Domed | |
Yes | |
Autoclavable |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title