Learn More
BrandTech™ BRAND™ PCR 12-Strip Caps
Manufactured from polypropyle without lubricants or chemical additives that can contaminate reagents and samples.PCR 12-Strip Caps are autoclavable and RNase-, DNase-, and DNA-free. Caps are domed and available in a variety of colors for sample identification.
Supplier: BrandTech™ BRAND™ 781292
Description
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples.- Autoclavable
- RNase-, DNase-, and DNA-free
- All 12-Strip caps are domed and available in a variety of colors for sample identification
Specifications
Yellow | |
Domed | |
Yes | |
Autoclavable |
PCR tubes | |
Polypropylene | |
Yes | |
Tube Strip Cap |