Learn More
Labconco™ Paramount™ Ductless Enclosure Accessory, Carbon Filter
SDPSCI_ED
Labconco™ Paramount™ Ductless Enclosure Accessory, Carbon Filter used with Paramount Ductless Enclosures.
Supplier:  Labconco™ 6938200/EMD
 View more versions of this product
Catalog No. S28166
Add to Cart
Description
Description
  • Used with carbon-based and HEPA filters in variety of Labconco enclosures which require periodic replacement
  • Select the right filters to fit your Labconco product
Order Info
Acid gas detector tubes available. (See Cat. No. 10-030-241)
Specifications
Specifications
Carbon
1.5 in.
4.5 Kg
15 in.
3.8 cm
25.4 to 50.8 cm
Filter
Paramount Ductless Enclosure
10 lb.
10 to 20 in.
18 in.
38.1 cm
45.7 cm
Organic-vapor
Product Suggestions
Product Suggestions