Learn More
VistaLab™ Technologies Ovation M PROMO
SDP
The Ovation M is the ideal ergonomic, manual pipette that is autoclavable for easy sterilization.
$262.50 - $942.00
Specifications
Description Mechanical volume adjust pipette
Product Line Ovation™ M
View More Specs
Products 7
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
10-101-118TI
SDP
View Documents
VistaLab™ Technologies
10700010TI
1 to 10 mL
Each for $262.50
Add to Cart
10-101-119TI
SDP
View Documents
VistaLab™ Technologies
10700020TI
2 to 20 mL
Each for $262.50
Add to Cart
10-101-120TI
SDP
View Documents
VistaLab™ Technologies
10700100TI
10 to 100 µL
Each for $262.50
Add to Cart
10-101-121TI
SDP
View Documents
VistaLab™ Technologies
10700200TI
20 to 200 µL
Each for $262.50
Add to Cart
10-101-124TI
SDP
View Documents
VistaLab™ Technologies
10701000TI
100 to 1000 µL
Each for $262.50
Add to Cart
10-101-123TI
SDP
View Documents
VistaLab™ Technologies
10700999TI
0.2 to 2 µL, 2 to 20 µL, 20 to 200 µL, 100 to 1000 µL
Each for $942.00
Add to Cart
10-101-122TI
SDP
View Documents
VistaLab™ Technologies
10700998TI
1 to 10 µL, 10 to 100 µL, 2 to 200 µL
Each for $692.00
Add to Cart
Description
Description
  • Mechanical 2-stroke (overblow) low-force plunger operation
  • Mechanical precise click stop volume adjustment
  • Autoclavable for easy sterilization
  • Built-in microtube cap opener
  • Shelf hanger included with each pipette
  • Two-year warranty
Specifications
Specifications
Mechanical volume adjust pipette
Ovation™ M