Learn More
VistaLab™ Technologies Ovation M PROMO
The Ovation M is the ideal ergonomic, manual pipette that is autoclavable for easy sterilization.
Supplier: VistaLab™ Technologies 10700998TI
Description
- Mechanical 2-stroke (overblow) low-force plunger operation
- Mechanical precise click stop volume adjustment
- Autoclavable for easy sterilization
- Built-in microtube cap opener
- Shelf hanger included with each pipette
- Two-year warranty
Specifications
Yes | |
Mechanical volume adjust pipette | |
Ovation™ M | |
1 to 10, 10 to 100, 2 to 200 μL |