Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Ovation™ 8-Channel Pipetters PROMO
SDP

Straightforward, simple programming coupled with reliable, consistent accuracy and precision.

Supplier:  VistaLab™ Technologies 11600125TI

 View more versions of this product

Catalog No. 22-146-676TI


Add to Cart

Description

Description

  • Ergonomically correct, comfort-driven design with electronic functions for repetitive procedures
  • Feature a tip acquisition system that provides a positive feedback signal to indicate every tip is securely seated and sealed; no manual manipulations needed
  • Upon completion of pipetting, all tips are discarded with the push of a button
  • Equipped with Multiple Dispense, Pipet, Reverse Pipet, Mix and Serial Dilute operations and liquid handling functions with recall of stored volume settings
  • Available in both left- and right-handed models with suitably placed LCD screen
  • For precision liquid dispensing from 0.5 to 1250μL

Specifications

Specifications

±0.8% at 125 μL, ±1% at 62.5 μL, ±3% (at 12.5 μL)
Right Handed
Technologies Ovation™
2 to 125 μL
Ergonomic
1 Year
Small tip
8
Electronic Pipette
LCD
0.5 μL
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit