Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORP Standard, 250 mV vs. Ag/AgCl, Reagents

ORP Standard, 250 mV vs. Ag/AgCl, Reagent, use for Redox. Recommended Storage: 2°C to 8°C (36°F to 46°F)Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material ORP Standard, 250 mV vs. Ag/AgCl
CAS 7732-18-5, 7447-40-7, 14459-95-1
Packaging Opaque HDPE
Color Yellow/Green
For Use With (Equipment) Redox Meter
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-334-584
View Documents
Reagents Holdings Llc
CO0002504BP
4 L
Each for $392.76
N/A
Due to product restrictions, please to purchase or view availability for this product.
01-334-583
View Documents
Reagents Holdings Llc
CO000250500B
500 mL
Each for $93.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Specifications

Specifications

ORP Standard, 250 mV vs. Ag/AgCl
Opaque HDPE
Redox Meter
Reagent
1.0052
Completely miscible
7732-18-5, 7447-40-7, 14459-95-1
Yellow/Green
Oxidation Reduction Potential Standard
100°C
Redox
Liquid
SDS
Documents

Documents

Product Certifications
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit