Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORP Standard, 250 mV vs. Ag/AgCl, Reagents
ORP Standard, 250 mV vs. Ag/AgCl, Reagent, use for Redox. Recommended Storage: 2°C to 8°C (36°F to 46°F)Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CO000250500B
Specifications
ORP Standard, 250 mV vs. Ag/AgCl | |
7732-18-5, 7447-40-7, 14459-95-1 | |
Yellow/Green | |
Oxidation Reduction Potential Standard | |
100°C | |
Redox | |
Liquid |
Opaque HDPE | |
500 mL | |
Redox Meter | |
Reagent | |
1.0052 | |
Completely miscible |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction