Learn More
MilliporeSigma™ Supelco™ ORBO™ 80 Isocyanate Filter, No Cassettes
Used for gas and vapor sampling
Supplier: MilliporeSigma™ Supelco™ 20811
Description
ORBO™ 80 Coated 1,2-PP Filter is a 1-(2-pyridyl) piperazine coated glass fiber filter best suited for sampling diisocyanates in the air (OSHA Methods 42 and 47).Specifications
No cassettes | |
Glass | |
25/Pk. | |
37 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
ORBO is a trademark of Sigma-Aldrich Co. LLC