Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ BamHI, Fisher BioReagents™

 View more versions of this product

Catalog No. BP8005-5

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

5'...G^G A T C C...3'
3'...C C T A G^G...5'

Conditions for 100% Activity:

  • 1X OPTIZYME Buffer BamHI: 10mM Tris-HCl (pH 8.0 at 37°C), 5mM MgCl2, 100mM KCl, 0.02% Triton X-100 and 0.1mg/ml BSA.
  • Incubate at 37°C
Supplied with: 10X OPTIZYME BamHI Buffer
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 20 - 50%™
  • Buffer 2: 100%
  • Buffer 3: 20 - 50%
  • Buffer 4: 100%™
  • Buffer 5: 50 - 100%™
™star activity occurs at greater than 5-fold overdigestion (5 units × 1 hour)
Storage Buffer:
10mM Tris-HCl (pH 7.4 at 25°C), 200mM NaCl, 1mM DTT, 0.1mM EDTA, 0.15% Triton X-100, 0.2mg/ml BSA and 50% (v/v) glycerol.
Ligation and Recleavage:
More than 95% of DNA fragments can be ligated and recut after a 50-fold overdigestion with BamHI.
Digestion of Agarose-embedded DNA:
A minimum of 5 units of BamHI is required for the complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
Note: Low salt, high glycerol (>5%) concentrations, pH >8.0 or a large excess of enzyme may result in star activity.
Compatible Ends: BclI, BglII, Bsp143I, MboI, PsuI.
Methylation Effects:
Dam: completely overlaps - no effect.
Dcm: may overlap - no effect.
CpG: may overlap - no effect.
EcoKI: never overlaps - no effect.
EcoBI: never overlaps - no effect.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 U/μL
37°C
>95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with BamHI
Keep container tightly closed
G.GATCC
Liquid
25 to 50% Water, >50% Glycerin, 1 to 2.5% Sodium Chloride
7.4
10mM Tris HCl (pH 7.4 at 25°C), 200mM NaCl, 1mM DTT, 0.1mM EDTA, 0.15% Triton™ X-100, 0.2mg/mL BSA, 50% Glycerol
12,500 U
BamHI
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit