Learn More
Dunlop™ Onguard™ Men's Flex 3 Work Boots
Supplier: Dunlop™ 89908080
This item is not returnable.
View return policy
Specifications
ASTM F2413-05 | |
Pull-on | |
16 in. | |
PVC, Nitrile | |
8 | |
Steel | |
Yes |
Black | |
Used in oil rigs, refineries, food processing, general industrial areas, lawn care, dairies, breweries and utilities | |
Kick-off | |
Steel | |
Gray/yellow power-lug outsole and removable air cushion insole, slip-resistant | |
Fats, oils and chemicals |