Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dunlop™ Onguard™ Goliath™ PVC Knee Boots
Ideal for general industrial and safety applications
$93.00 - $113.10
Specifications
Certifications/Compliance | ASTM F2413-05 |
---|---|
Color | Black |
Design | Pull-on |
For Use With (Application) | Used in food processing, chemical plants, lawn care, canneries, poultry processing, fish processing, breweries and mining |
Height (English) | 16 in. |
Catalog Number | Mfr. No. | Size | Toe Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Toe Style | Price | Quantity & Availability | ||||
19-099-168F
|
Dunlop™
89680 10 |
10 | Plain |
Pair for $99.60
|
|
||||
19-099-168J
|
Dunlop™
89680 13 |
13 | Plain |
Pair for $96.50
|
|
||||
19-099-168G
|
Dunlop™
89680 11 |
11 | Plain |
Pair for $96.50
|
|
||||
19-099-168H
|
Dunlop™
89680 12 |
12 | Plain |
Pair for $96.50
|
|
||||
19-099-168K
|
Dunlop™
89680 14 |
14 | Plain |
Pair for $96.00
|
|
||||
19-099-168B
|
Dunlop™
89680 6 |
6 | Plain |
Pair for $96.50
|
|
||||
19-099-168C
|
Dunlop™
89680 7 |
7 | Plain |
Pair for $99.60
|
|
||||
19-099-168D
|
Dunlop™
89680 8 |
8 | Plain |
Pair for $93.00
|
|
||||
19-099-167F
|
Dunlop™
89682 10 |
10 | Safety Steel Toe |
Pair for $113.10
|
|
||||
19-099-168E
|
Dunlop™
89680 9 |
9 | Plain |
Pair for $96.00
|
|
||||
19-099-167J
|
Dunlop™
89682 13 |
13 | Safety Steel Toe |
Pair for $113.10
|
|
||||
19-099-167B
|
Dunlop™
89682 6 |
6 | Safety Steel Toe |
Pair for $108.90
|
|
||||
19-099-167K
|
Dunlop™
89682 14 |
14 | Safety Steel Toe |
Pair for $108.90
|
|
||||
19-099-167C
|
Dunlop™
89682 7 |
7 | Safety Steel Toe |
Pair for $105.00
|
|
||||
19-099-167D
|
Dunlop™
89682 8 |
8 | Safety Steel Toe |
Pair for $108.90
|
|
||||
19-099-167G
|
Dunlop™
89682 11 |
11 | Safety Steel Toe | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-099-167H
|
Dunlop™
89682 12 |
12 | Safety Steel Toe | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
19-099-167E
|
Dunlop™
89682 9 |
9 | Safety Steel Toe | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Constructed of a special urethane/PVC compound with Elvaloy™ and polymeric plasticizers for enhanced strength and high abrasion resistance
- Superior protection from animal fats (especially poultry), acids, oils, greases, and other chemicals
- Net-lined for easy on and off, with a removable insole
- Built-in steel shank for firm arch support
- Black with a gray Power-Lug outsole
Order Info
Available with or without steel toe in men's sizes only
Compliance
Steel toe meets or exceeds ASTM™ F2413-11 specifications.
Specifications
ASTM F2413-05 | |
Pull-on | |
16 in. | |
Steel | |
Animal fats, acids, oils, greases and chemicals |
Black | |
Used in food processing, chemical plants, lawn care, canneries, poultry processing, fish processing, breweries and mining | |
Urethane, PVC | |
Gray power-lug outsole and removable insole | |
Yes |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title