Learn More
Dunlop™ Onguard™ Goliath™ PVC Knee Boots
SDP
Ideal for general industrial and safety applications
Supplier:  Dunlop™ 89680 9
 View more versions of this product
Catalog No. 19-099-168E
Explore available promotions
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Constructed of a special urethane/PVC compound with Elvaloy™ and polymeric plasticizers for enhanced strength and high abrasion resistance
  • Superior protection from animal fats (especially poultry), acids, oils, greases, and other chemicals
  • Net-lined for easy on and off, with a removable insole
  • Built-in steel shank for firm arch support
  • Black with a gray Power-Lug outsole
Order Info
Available with or without steel toe in men's sizes only
Compliance
Steel toe meets or exceeds ASTM™ F2413-11 specifications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ASTM F2413-05
Pull-on
16 in.
Steel
Gray power-lug outsole and removable insole
Animal fats, acids, oils, greases and chemicals
Black
Used in food processing, chemical plants, lawn care, canneries, poultry processing, fish processing, breweries and mining
Urethane, PVC
9
Plain
Yes
Product Suggestions
Product Suggestions