Learn More
Diba Omnifit™ Replacement Filters for Bottom-of-Bottle Filter from Cole-Parmer™
Diba™ Omnifit™ Replacement Filters for Bottom-of-Bottle Filter from Cole-Parmer™ are designed to maintain the cleanest possible solvent supply.
Catalog Number | Mfr. No. | Description | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | For Use With (Application) | Price | Quantity & Availability | ||||
OF9FE3210PE
|
Diba
2194337 |
GL32/38-430 | Bottom-of-bottle filter 009F-32-10-PE | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
OF9FE4510PE
|
Diba
2194339 |
GL45 | Bottom-of-bottle filter 009F-45-10-PE | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Use with bottom-of-bottle filter
- Made from PE
- 20/pk
Specifications
Polyethylene | |
10 μm | |
Replacement Filter |