Learn More
Diba Omnifit™ Replacement Filters for Bottom-of-Bottle Filter from Cole-Parmer™
Diba™ Omnifit™ Replacement Filters for Bottom-of-Bottle Filter from Cole-Parmer™ are designed to maintain the cleanest possible solvent supply.
Supplier: Diba 2194339
Description
- Use with bottom-of-bottle filter
- Made from PE
- 20/pk
Specifications
GL45 | |
10 μm | |
Replacement Filter |
Polyethylene | |
Bottom-of-bottle filter 009F-45-10-PE |