Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purus™ Non-Latex CLEAN ImagePremier Cleanroom Paper: PROMO

Suitable for use in Class 10 to Class 1000 cleanrooms

Supplier:  Purus™ PCIB1084CPM

 View more versions of this product

Catalog No. 19-125-892PM

Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.


Description

Description

  • Made of polyethylene
  • Binding technology reduces paper fiber
  • Cleaned and double bagged in class 10 cleanroom
  • Produced in Class 100 cleanroom
  • Double sided web cleaned prior to sheeting
  • Printability and heat toner adhesion
Specifications

Specifications

Polyethylene
8.5 x 11 in.
279 mm
216 mm
Laser printers and copy machines
22 lb.
Blue
11 in.
8.5 in.
For Class 10 to Class 1000 cleanrooms, writing, printing and photocopying in clean environments
Cleanroom Paper
9.97 kg
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit