Learn More
KleenGuard™ Nemesis™ Safety Glasses
![GREENER_CHOICE](/content/dam/fishersci/glyphs/greener_choice.png)
The foam safety eyewear offers stylish looks, durability, and lightweight design for protection at work and home. With polycarbonate lenses providing 99.9% UVA/UVB/UVC protection, it prevents eye damage. The wraparound frame offers peripheral protection, soft touch temples ensure comfort, and the flexible nose piece prevents slippage. It meets ANSI/ISEA Z87.1+ standards.
Supplier: KleenGuard™ 65336
This item is not returnable.
View return policy
Description
- Anti-Fog Coating
- Clear Lenses
- Black Frame
- Unisex for Men and Women
- Detachable and re-attachable foam gasket provides comfort and helps to seal out dust and debris
- KleenVision™ Anti-Fog Coating- ideal for humid environments and temperature variances
- Recyclable with RightCycle™ by Kimberly-Clark Professional™
- Meets ANSI Z87.1+ 2020 standards for high impact and anti-fog
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Anti-fog | |
Safety Glasses | |
Reusable | |
99.9% UVA/UVB/UVC protection |
Black | |
Black | |
Polycarbonate |