Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Danger Safety Glasses Required Beyond This Point Signs
$21.20 - $31.15
Specifications
Color | Black/Red on White |
---|---|
Message | DANGER Safety Glasses Required Beyond This Point |
Certifications/Compliance | OSHA |
Quantity | 1 Ea. |
For Use With (Application) | Alert employees/guests of hazards |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
17-985-44C
|
National Marker™
D 108 R |
Plastic |
Each for $31.15
|
|
17-985-44A
|
National Marker™
D 108 P |
Vinyl |
Each for $21.20
|
|
Specifications
Black/Red on White | |
OSHA | |
Alert employees/guests of hazards | |
Safety Sign |
DANGER Safety Glasses Required Beyond This Point | |
1 Ea. | |
7 in. | |
10 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title