Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Danger Safety Glasses Required Beyond This Point Signs
Supplier: National Marker™ D 108 P
Specifications
OSHA | |
1 Ea. | |
Alert employees/guests of hazards | |
178°F | |
0.004 in. | |
DANGER Safety Glasses Required Beyond This Point | |
Red on White |
Vinyl | |
Surface Mount | |
7 in. | |
81°C | |
10 in. | |
Safety Sign |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction