Learn More
Thermo Scientific™ National Assembled Convenience Kits: EPA Vials with Caps and Septa
Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.
Catalog Number | Mfr. No. | Volume (Metric) | Color | Certifications/Compliance | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Color | Certifications/Compliance | Price | Quantity & Availability | |||
03-377-33
|
Thermo Scientific™
B7920C |
20 mL | Clear | Class 200 |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||
03-377-34
|
Thermo Scientific™
B7920C3 |
20 mL | Clear | Class 300 |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||
03-377-302
|
Thermo Scientific™
B7950C |
40 mL | Clear | Class 200 |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||
03-377-41
|
Thermo Scientific™
B7951C |
40 mL | Amber | Class 200 |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||
03-377-304
|
Thermo Scientific™
B7951C3 |
40 mL | Amber | Class 300 |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||
Description
Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.
- Recommended for discrete water sampling under EPA 40 CFR 136 “Guidelines for Establishing Test Procedures for the Analysis of Pollutants” and EPA 40 CFR 141 “National Interim Primary Drinking Water Regulations: Control of Trihalomethanes in Drinking Water”
- Class 100—Vials, Caps and Septa are assembled
- Class 200—Vials, Caps and Septa are processed to EPA specifications and assembled
- Class 300—Vials, Caps and Septa are processed and certified to EPA specifications, then assembled (Certificate of Analysis included with each box of vials)
Specifications
Discrete Water Sampling Applications |