Learn More
Thermo Scientific™ National Assembled Convenience Kits: EPA Vials with Caps and Septa
Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.
Supplier: Thermo Scientific™ B7950C
Description
Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.
- Recommended for discrete water sampling under EPA 40 CFR 136 “Guidelines for Establishing Test Procedures for the Analysis of Pollutants” and EPA 40 CFR 141 “National Interim Primary Drinking Water Regulations: Control of Trihalomethanes in Drinking Water”
- Class 100—Vials, Caps and Septa are assembled
- Class 200—Vials, Caps and Septa are processed to EPA specifications and assembled
- Class 300—Vials, Caps and Septa are processed and certified to EPA specifications, then assembled
Includes:
Certificate of Analysis is included with each box of vials.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Clear; 40mL | |
40 mL | |
24-400 |
Clear | |
Class 200 | |
Discrete Water Sampling Applications |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.