Learn More
Mystaire™ Ductless Forensic Workstation Pre-filter
For use with the Mystaire Ductless Forensic Workstation (FE-36). An electrostatically charged fiber filter which effectively removes submicron particulates and is designed to protect the adsorption filter from contaminants such as smoke, acid mists, and other submicron particulates.
Supplier: Mystaire™ FE36PF
Description
- Made in the U.S.A.
- Each filter application is reviewed to ensure a proper filtration package is selected
- Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)