Learn More
MSA™ Airline Filter
High-efficiency filtration of compressed breathing air
Supplier:  MSA™ 81857
 View more versions of this product
Catalog No. 19-041-913
Add to Cart
Includes: One particulate filter and two organic vapor cartridges. Replacement Kit includes one particulate filter, two organic vapor cartridges, O-ring, three gasket cartridges, and a gasket retainer.
Description
Description
  • Designed especially for protection against dust, mists, fumes, smoke, and petroleum vapors
  • Removes at least 99% of 0.3μm and larger particulates when used with constant flow and pressure-demand airline respirators
  • Filter can be used at inlet pressures up to 125psig (8618hPag), with a pressure drop of 1psig (69hPag) at a maximum rated airflow of 25 cu. ft./min. (0.7m3/min.)
  • Inlet and Outlet Fittings: 1/2 in. (1.3cm) NPT female
Does not remove carbon monoxide. Cartridges should be replaced after 1000 hours of use, or sooner if odor is detectable or pressure drop increases.
Order Info
Weight: 3 lb. (1.4kg)
Specifications
Specifications
NIOSH
Filtration of the Airstream™ after it leaves the compressor, 99 percent removal of 0.3μ and larger particulates including dusts, mists, fumes, smokes, petroleum vapors and organic vapors
1 Ea.
0.5 in. NPT female
Airline Filter
Female Fittings
Product Suggestions
Product Suggestions