Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Hormones: Dexamethasone
Used to study apoptosis, cell signaling pathways and gene expression. MP Biomedicals Hormones: Dexamethasone acts as anti-inflammatory glucocorticoid that induces the production of phospholipase A2 inhibitory protein (lipocortin).
$73.00 - $670.00
Specifications
Content And Storage | 15° to 30°C |
---|---|
Form | Crystalline powder |
Product Type | Dexamethasone |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ICN19456125
|
MP Biomedicals™
0219456125 |
25 mg |
Each for $73.00
|
|
|||||
ICN19456180
|
MP Biomedicals™
0219456180 |
100 mg |
Each for $162.00
|
|
|||||
ICN19456190
|
MP Biomedicals™
0219456190 |
500 mg |
Each for $453.00
|
|
|||||
ICN19456191
|
MP Biomedicals™
0219456191 |
1000 mg |
Each for $670.00
|
|
|||||
Description
- Inhibits induction of nitric oxide synthase (IC50=5 nM)
- Causes reduction in cyclin A and Cdk2 activity, inhibition of G1/S transition in osteoblasts and inhibition of phosphorylation of Rb protein
- Observed to induce apoptosis in human thymocytes and eosinophils but to inhibit apoptosis in neutrophils
Specifications
15° to 30°C | |
Crystalline powder | |
Dexamethasone |
For research use only. Not intended for therapeutic use.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title