Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hormones: Dexamethasone

Used to study apoptosis, cell signaling pathways and gene expression. MP Biomedicals Hormones: Dexamethasone acts as anti-inflammatory glucocorticoid that induces the production of phospholipase A2 inhibitory protein (lipocortin).

Supplier:  MP Biomedicals™ 0219456190

 View more versions of this product

Catalog No. ICN19456190


Add to Cart

Description

Description

  • Inhibits induction of nitric oxide synthase (IC50=5 nM)
  • Causes reduction in cyclin A and Cdk2 activity, inhibition of G1/S transition in osteoblasts and inhibition of phosphorylation of Rb protein
  • Observed to induce apoptosis in human thymocytes and eosinophils but to inhibit apoptosis in neutrophils
Specifications

Specifications

15° to 30°C
Crystalline powder
Dexamethasone
500 mg
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For research use only. Not intended for therapeutic use.