Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Metric Measuring Spoon Set



Unbreakable plastic set of five metric measuring spoons.
Supplier: United Scientific™ UNSPOONB
Description
- Unbreakable plastic set of five metric measuring spoons
- Includes the following: 1.25 mL (1/4 tsp), 2.5 mL (1/2 tsp), 5 mL (1 tsp), 7.5 mL (1/2 Tbsp) and 15 mL (1 Tbsp)
Specifications
Measuring Spoon | |
Not Autoclavable | |
1.25 mL, 2.5 mL, 5 mL, 7.5 mL, 15 mL | |
Measuring Small Samples |
Plastic | |
1/4 tsp, 1/2 tsp, 1 tsp, 1/2 Tbsp, 1 Tbsp | |
Non-sterile |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title