Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medegen Biohazardous Waste Bags, HDPE Film, Flat Pack
Passes ASTMD1709-85, 165 Gram Dart Test
$251.00 - $300.00
Specifications
Product Type | Biohazardous Waste Bag |
---|---|
Certifications/Compliance | ASTM Dart |
Color | Red, Black |
For Use With (Application) | Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab |
Includes | Biohazardous or infectious waste with biohazard symbol |
Catalog Number | Mfr. No. | Dimensions (L x W) | Thickness (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Thickness (Metric) | Price | Quantity & Availability | ||||
14-828-190
|
Medegen
HRD404816 |
40 x 48 in. (101.6 x 121.92cm) | 16 micron |
Case of 250 for $251.00
|
|
||||
14-828-188
|
Medegen
HRD334014 |
33 x 40 in. (83.82 x 101.6cm) | 14 micron |
Case of 500 for $300.00
|
|
||||
14-828-187
|
Medegen
HRD304314 |
30 x 43 in. (76.2 x 109.22cm) | 14 micron |
Case of 500 for $273.00
|
|
||||
14-828-189
|
Medegen
HRD384616 |
38 x 46 in. (96.52 x 116.84cm) | 16 micron | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Made from HDPE
- Minimize the risk of contamination from biohazardous materials during collection and transport
- Bags are red with “Biohazardous Waste/Infectious Waste” and biohazard symbol imprinted in black ink
- Individually folded
Specifications
Biohazardous Waste Bag | |
Red, Black | |
Biohazardous or infectious waste with biohazard symbol |
ASTM Dart | |
Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab | |
HDPE |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title