Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medegen Biohazardous Waste Bags, HDPE Film, Flat Pack
Passes ASTMD1709-85, 165 Gram Dart Test
Supplier: Medegen HRD334014
Description
- Made from HDPE
- Minimize the risk of contamination from biohazardous materials during collection and transport
- Bags are red with “Biohazardous Waste/Infectious Waste” and biohazard symbol imprinted in black ink
- Individually folded
Specifications
Biohazardous Waste Bag | |
ASTM Dart | |
Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab | |
HDPE | |
33 x 40 in. (83.82 x 101.6cm) | |
83.82 cm | |
101.6 cm |
33 gal. | |
Red, Black | |
Biohazardous or infectious waste with biohazard symbol | |
14 micron | |
33 in. | |
40 in. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction