Learn More
Laxco™ Mechanical Stage Upgrade Kit
This Mechanical Stage Upgrade Kit converts a fixed stage to a mechanical stage. Options for SLi3 PRO Series and SLi6 PRO Series Microscopes.
$915.00 - $1566.00
Specifications
Warranty | 5 years optical and mechanical, 1 year electrical from date of purchase |
---|---|
Stage | Low Position Coaxial Mechanical Stage |
Product Line | SLi3 PRO Series |
Product Type | Mechanical Stage |
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
SLI3P-ME-KIT2
|
Laxco™
SLI3PMEKIT2 |
SLi3 PRO Series Microscopes |
Each for $915.00
|
|
|||||
SLI6P-ME-KIT1
|
Laxco™
SLI6PMEKIT1 |
SLi6 PRO Series Microscopes |
Each for $1,566.00
|
|
|||||
Description
- Low position coaxial mechanical stage
- Includes stage inserts (petri dish, slide, terasaki) and stage extension
- Options available for use with SLi3 PRO Series Microscopes or SLi6 PRO Series Microscopes
Specifications
5 years optical and mechanical, 1 year electrical from date of purchase | |
Low Position Coaxial Mechanical Stage | |
SLi3 PRO Series | |
Mechanical Stage |