Learn More
Laxco™ Mechanical Stage Upgrade Kit
This Mechanical Stage Upgrade Kit converts a fixed stage to a mechanical stage. Options for SLi3 PRO Series and SLi6 PRO Series Microscopes.
Supplier: Laxco™ SLI3PMEKIT2
Description
- Low position coaxial mechanical stage
- Includes stage inserts (petri dish, slide, terasaki) and stage extension
- Options available for use with SLi3 PRO Series Microscopes or SLi6 PRO Series Microscopes
Specifications
SLi3 PRO Series Microscopes | |
Low Position Coaxial Mechanical Stage | |
SLi3 PRO Series |
5 years optical and mechanical, 1 year electrical from date of purchase | |
Moving Range of 120 x 78 mm | |
Mechanical Stage |