Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ MALP-2, TLR6 and TLR2 ligand
Applications: Functional, In vitro assay
$723.50
Specifications
Concentration | 0.1 mg/mL |
---|---|
Formulation | C99H167N19O30S |
Content And Storage | Store at -20C. Avoid freeze-thaw cycles. |
Target | MALP-2, TLR6 and TLR2 Ligand |
Species | Human/Mouse/Avian |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP226219
|
Novus Biologicals™
NBP226219 |
50 μg |
Each for $723.50
|
|
|||||
NBP2262192
|
Novus Biologicals™
NBP2262192UG |
2μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Specific Activity is approx. 2x10^8units/mg. One unit is defined as the dilution giving half maximal release of nitric oxide from C3H/HeJ mouse peritoneal exudate cells in the standard assay. Immunogen: MALP-2, TLR6 and TLR2 ligand Buffer: 2μg in 20μL of sterile PBS solution containing 2.5% (v/v) 2-propanol, 25mM n-octyl-b-D-glucopyranoside and 1% (wt/v) human serum albumin. Dilution: Functional, In vitro assay MALP-2 was originally isolated from Mycoplasma fermentans. This MALP-2 corresponds to the originally isolated isomer, which expresses potent endotoxin-like activity and approaches in certain experimental systems the toxicity of LPS. MW: 2135.2Specifications
0.1 mg/mL | |
Store at -20C. Avoid freeze-thaw cycles. | |
Human/Mouse/Avian |
C99H167N19O30S | |
MALP-2, TLR6 and TLR2 Ligand |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title