Learn More
Simport™ Scientific M480 Disposable Embedding Cassettes in Quickload™ Stacks
M480 Disposable Embedding Cassettes in Quickload™ Stacks for holding and identifying tissue samples in processing/embedding procedures.
Supplier:  Simport™ Scientific M4803T
 View more versions of this product
Catalog No. 22-038-035
Add to Cart
Description
Description
  • In Quickload™ stacks
  • 45° Anterior writing area
  • Accept standard metal lids (cat# M481)
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Specifications
2000/Pk.
Pink
Acetal